2-Amino-4-piperidino-6-methylpyrimidine ≥95%

Supplier: Matrix Scientific

073677-1G 073677-5G 073677-10G
102531-428EA 533.85 USD
102531-428 102531-430 102531-432
2-Amino-4-piperidino-6-methylpyrimidine ≥95%
2-Amino-4-piperidino-6-methylpyrimidine

Hazard: Irritant

Formula: C₁₀H₁₆N₄
MW: 192.26 g/mol
MDL Number: MFCD01011781
CAS Number: 91717-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR