5-Bromo-1H-pyrrolo[3,2-b]pyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-4-azaindole

073675-250MG 073675-1G 073675-5G
102531-418EA 541.54 USD
102531-418 102531-420 102531-422
5-Bromo-1H-pyrrolo[3,2-b]pyridine ≥95%
5-Bromo-1H-pyrrolo[3,2-b]pyridine

Hazard: Irritant

Formula: C₇H₅BrN₂
MW: 197.0300 g/mol
MDL Number: MFCD09263244
CAS Number: 1000341-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR