7-Chloro-1H-pyrrolo[3,2-b]pyridine ≥95%

Supplier: Matrix Scientific

073674-250MG 073674-1G
102531-414EA 967.69 USD
102531-414 102531-416
7-Chloro-1H-pyrrolo[3,2-b]pyridine ≥95%
7-Chloro-1H-pyrrolo[3,2-b]pyridine

Hazard: Irritant

Formula: C₇H₅ClN₂
MW: 152.5800 g/mol
MDL Number: MFCD08234945
CAS Number: 357263-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR