5-Aminolevulinic acid hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Amino-4-ketovaleric acid hydrochloride, 5-Amino-4-oxopentanoic acid hydrochloride, δ-Aminolevulinic Acid Hydrochloride

073673-1G 073673-5G 073673-10G
102531-408EA 38.46 USD
102531-408 102531-410 102531-412
5-Aminolevulinic acid hydrochloride ≥95%
5-Aminolevulinic acid hydrochloride

Hazard: Irritant

Formula: C₅H₁₀ClNO₃
MW: 167.59 g/mol
Melting Pt: 145 °C
MDL Number: MFCD00012869
CAS Number: 5451-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR