1-Boc-3-iodopiperidine ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 3-iodopiperidine-1-carboxylate

073672-250MG 073672-1G 073672-5G
102531-402EA 133.85 USD
102531-402 102531-404 102531-406
1-Boc-3-iodopiperidine ≥95%
1-Boc-3-iodopiperidine

Hazard: Irritant

Formula: C₁₀H₁₈INO₂
MW: 311.1599 g/mol
MDL Number: MFCD08059309
CAS Number: 850761-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR