6-Bromo-2-methylisoquinolin-1(2H)-one ≥95%

Supplier: Matrix Scientific

073671-250MG 073671-1G 073671-5G
102531-396EA 640 USD
102531-396 102531-398 102531-400
6-Bromo-2-methylisoquinolin-1(2H)-one ≥95%
6-Bromo-2-methylisoquinolin-1(2H)-one

Hazard: Irritant

Formula: C₁₀H₈BrNO
MDL Number: MFCD11858430
CAS Number: 864866-92-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR