6-Bromo-1,2,3,4-tetrahydroisoquinoline hydrochloride ≥95%
Supplier: Matrix Scientific
073669-250MG
073669-1G
073669-5G
102531-386EA
218.46
USD
102531-386
102531-388
102531-390
6-Bromo-1,2,3,4-tetrahydroisoquinoline hydrochloride ≥95%
6-Bromo-1,2,3,4-tetrahydroisoquinoline hydrochloride
Hazard: Irritant
Formula:
C₉H₁₁BrClN MW: 248.5473 g/mol |
MDL Number:
MFCD08234662 CAS Number: 215798-19-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...