1-Boc-(3-carboxymethoxy)azetidine ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Boc-(3-Carboxymethoxy)azetidine

073668-1G 073668-5G
102531-382EA 640 USD
102531-382 102531-384
1-Boc-(3-carboxymethoxy)azetidine ≥95%
1-Boc-(3-carboxymethoxy)azetidine

Hazard: Irritant

Formula: C₁₀H₁₇NO₅
MW: 231.250 g/mol
MDL Number: MFCD02179034
CAS Number: 889952-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR