(S)-2,4-Diaminobutanoic acid dihydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
L-2,4-Diaminobutanoic acid dihydrochloride, H-Dab-OH 2HCl, L-2,4-Diaminobutyric acid dihydrochloride, (2S)-2,4-Diaminobutanoic acid dihydrochloride, H-Daba-OH·2HCl, (S)-(+)-2,4-Diaminobutyric acid dihydrochloride
073667-1G
073667-5G
102531-376EA
49.23
USD
102531-376
102531-378
(S)-2,4-Diaminobutanoic acid dihydrochloride ≥95%
(S)-2,4-Diaminobutanoic acid dihydrochloride
Hazard: Irritant
Formula:
C₄H₁₂Cl₂N₂O₂ MW: 191.06 g/mol |
MDL Number:
MFCD00064561 CAS Number: 1883-09-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...