6-Chloropyrazin-2-ol ≥95%

Supplier: Matrix Scientific

073665-250MG 073665-1G 073665-5G
102531-364EA 170.77 USD
102531-364 102531-366 102531-368
6-Chloropyrazin-2-ol ≥95%
6-Chloropyrazin-2-ol

Hazard: Irritant/Stored under Argon

Formula: C₄H₃ClN₂O
MW: 130.5300 g/mol
MDL Number: MFCD00204104
CAS Number: 4925-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR