3-Amino-6-chloropyrazine-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

073664-250MG 073664-1G 073664-5G
102531-358EA 498.46 USD
102531-358 102531-360 102531-362
3-Amino-6-chloropyrazine-2-carboxylic acid ≥95%
3-Amino-6-chloropyrazine-2-carboxylic acid

Hazard: Irritant

Formula: C₅H₄ClN₃O₂
MW: 173.5600 g/mol
MDL Number: MFCD10697761
CAS Number: 2727-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR