6-Chloroquinoline-8-carboxylic acid ≥95%

Supplier: Matrix Scientific

073663-1G 073663-5G 073663-10G
102531-352EA 533.85 USD
102531-352 102531-354 102531-356
6-Chloroquinoline-8-carboxylic acid ≥95%
6-Chloroquinoline-8-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₆ClNO₂
MDL Number: MFCD02752433
CAS Number: 6456-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR