4-Chloropyrimidin-5-amine ≥95%

Supplier: Matrix Scientific

073662-1G 073662-5G
102531-348EA 461.54 USD
102531-348 102531-350
4-Chloropyrimidin-5-amine ≥95%
4-Chloropyrimidin-5-amine

Hazard: Irritant

Formula: C₄H₄ClN₃
MDL Number: MFCD06412561
CAS Number: 54660-78-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR