2-Chloro-7-(trifluoromethyl)quinoxaline ≥95%

Supplier: Matrix Scientific

073661-250MG 073661-1G
102531-344EA 783.08 USD
102531-344 102531-346
2-Chloro-7-(trifluoromethyl)quinoxaline ≥95%
2-Chloro-7-(trifluoromethyl)quinoxaline

Hazard: Irritant

Formula: C₉H₄ClF₃N₂
MW: 232.59 g/mol
MDL Number: MFCD13193272
CAS Number: 883-94-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR