2-Bromoisonicotinonitrile ≥95%

Supplier: Matrix Scientific

073658-5G 073658-25G
102531-328EA 89.23 USD
102531-328 102833-648
2-Bromoisonicotinonitrile ≥95%
2-Bromoisonicotinonitrile

Hazard: Irritant

Formula: C₆H₃BrN₂
MW: 183.01 g/mol
Melting Pt: 70…74 °C
MDL Number: MFCD07367879
CAS Number: 10386-27-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR