2-Chloro-3-nitro-5-(trifluoromethyl)pyridine ≥95%

Supplier: Matrix Scientific

073657-1G 073657-5G 073657-25G
102531-320EA 1200 USD
102531-320 102531-322 102663-952
2-Chloro-3-nitro-5-(trifluoromethyl)pyridine ≥95%
2-Chloro-3-nitro-5-(trifluoromethyl)pyridine

Hazard: Irritant

Formula: C₆H₂ClF₃N₂O₂
MW: 226.54 g/mol
MDL Number: MFCD06255171
CAS Number: 72587-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR