2-Bromo-5-ethoxypyridin-3-ylamine ≥95%

Supplier: Matrix Scientific

073655-250MG 073655-1G 073655-5G
102531-310EA 355.38 USD
102531-310 102531-312 102531-314
2-Bromo-5-ethoxypyridin-3-ylamine ≥95%
2-Bromo-5-ethoxypyridin-3-ylamine

Hazard: Irritant

Formula: C₇H₉BrN₂O
MDL Number: MFCD03695819
CAS Number: 663193-84-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR