4-Methylpicolinic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Methylpicolinic acid, 4-Methylpyridine-2-carboxylic acid

073651-1G 073651-5G
102531-292EA 76.92 USD
102531-292 102531-294
4-Methylpicolinic acid ≥95%
4-Methylpicolinic acid

Hazard: Irritant

Formula: C₇H₇NO₂
MW: 137.14 g/mol
Melting Pt: 136-137°C
MDL Number: MFCD03731175
CAS Number: 4021-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR