6-Fluoro-2-methylnicotinic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Fluoro-2-methyl-3-pyridinecarboxylic acid

073650-1G 073650-5G
102531-286EA 267.69 USD
102531-286 102531-288
6-Fluoro-2-methylnicotinic acid ≥95%
6-Fluoro-2-methylnicotinic acid

Hazard: Irritant

Formula: C₇H₆FNO₂
MW: 155.1300 g/mol
MDL Number: MFCD05662400
CAS Number: 884494-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR