5-Bromo-6-trifluoromethylpyridin-2-ylamine ≥95%

Supplier: Matrix Scientific

073647-1G 073647-5G
102531-270EA 347.69 USD
102531-270 102531-272
5-Bromo-6-trifluoromethylpyridin-2-ylamine ≥95%
5-Bromo-6-trifluoromethylpyridin-2-ylamine

Hazard: Irritant

Formula: C₆H₄BrF₃N₂
MW: 241.0086 g/mol
MDL Number: MFCD11848010
CAS Number: 882500-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR