2-Chloro-5-fluoro-4-methylpyridine ≥95%

Supplier: Matrix Scientific

073646-1G 073646-5G
102531-262EA 53.85 USD
102531-262 102531-264
2-Chloro-5-fluoro-4-methylpyridine ≥95%
2-Chloro-5-fluoro-4-methylpyridine

Hazard: Irritant

Formula: C₆H₅ClFN
MDL Number: MFCD09864695
CAS Number: 881891-83-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR