6-Fluoroquinoline ≥95%

Supplier: Matrix Scientific

073644-1G 073644-5G 073644-10G
102531-252EA 66.15 USD
102531-252 102531-254 102531-256
6-Fluoroquinoline ≥95%
6-Fluoroquinoline

Hazard: Irritant

Formula: C₉H₆FN
MW: 147.15 g/mol
MDL Number: MFCD01685512
CAS Number: 396-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR