5-Methoxypyridine-3-boronic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 5-methoxypyridin-3-yl boronic acid, (5-Methoxypyridin-3-yl)boronic acid, 5-Methoxy-3-pyridineboronic acid

073643-1G 073643-5G
102531-248EA 347.69 USD
102531-248 102531-250
5-Methoxypyridine-3-boronic acid ≥95%
5-Methoxypyridine-3-boronic acid

Hazard: Irritant

Formula: C₆H₈BNO₃
MDL Number: MFCD03425977
CAS Number: 850991-69-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR