1-Boc-2-ethylpiperazine ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 2-ethylpiperazine-1-carboxylate

073642-1G 073642-5G
102531-244EA 569.23 USD
102531-244 102531-246
1-Boc-2-ethylpiperazine ≥95%
1-Boc-2-ethylpiperazine

Hazard: Irritant

Formula: C₁₁H₂₂N₂O₂
MW: 214.3100 g/mol
MDL Number: MFCD03265490
CAS Number: 393781-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR