tert-Butyl (4-aminocyclohexyl)carbamate ≥95%

Supplier: Matrix Scientific

Synonyms: N-Boc-1,4-diaminocyclohexane

073641-1G 073641-5G
102531-238EA 184.62 USD
102531-238 102531-240
tert-Butyl (4-aminocyclohexyl)carbamate ≥95%
tert-Butyl (4-aminocyclohexyl)carbamate

Hazard: Irritant

Formula: C₁₁H₂₂N₂O₂
MW: 214.31 g/mol
MDL Number: MFCD01076211
CAS Number: 195314-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR