(4-Methoxypyridin-2-yl)methanamine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(Aminomethyl)-4-methoxypyridine

073639-250MG 073639-1G 073639-5G
102531-228EA 456.92 USD
102531-228 102531-230 102531-232
(4-Methoxypyridin-2-yl)methanamine ≥95%
(4-Methoxypyridin-2-yl)methanamine

Hazard: Irritant

Formula: C₇H₁₀N₂O
MW: 138.1700 g/mol
MDL Number: MFCD11111886
CAS Number: 194658-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR