2-Fluoro-3-nitro-6-picoline ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Fluoro-3-nitro-6-methylpyridine, 2-Fluoro-6-methyl-3-nitropyridine

073638-1G 073638-5G 073638-10G
102531-222EA 92.31 USD
102531-222 102531-224 102531-226
2-Fluoro-3-nitro-6-picoline ≥95%
2-Fluoro-3-nitro-6-picoline

Hazard: Irritant

Formula: C₆H₅FN₂O₂
MW: 156.12 g/mol
MDL Number: MFCD03095067
CAS Number: 19346-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR