4-Amino-6-hydroxy-2-mercaptopyrimidine hydrate ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Amino-6-hydroxy-2-mercaptopyrimidine monohydrate, 4-Amino-6-hydroxy-2-mercaptopyrimidine Hydrate, 6-Amino-2-thiouracil hydrate, 6-Amino-2-thiouracil Hydrate

073637-1G 073637-5G 073637-10G
102531-216EA 143.08 USD
102531-216 102531-218 102531-220
4-Amino-6-hydroxy-2-mercaptopyrimidine hydrate ≥95%
4-Amino-6-hydroxy-2-mercaptopyrimidine hydrate

Hazard: Irritant

Formula: C₄H₅N₃OS·xH₂O
MW: 143.17 g/mol (anhy)
MDL Number: MFCD00052384
CAS Number: 65802-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR