5-Nitro-6-methyluracil ≥95%
Supplier: Matrix Scientific
073636-5G
073636-10G
073636-100G
102531-210EA
126.15
USD
102531-210
102531-212
102531-214
5-Nitro-6-methyluracil ≥95%
5-Nitro-6-methyluracil
Purity: 95+%
CAS: 16632-21-6
MDL Number: MFCD00047362
Molecular Formula: C5H5N3O4
Molecular Weight: 171,11
Hazard: Irritant
Formula:
C₅H₅N₃O₄ |
MDL Number:
MFCD00047362 CAS Number: 16632-21-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...