3-Methoxyisonicotinic acid ≥95%

Supplier: Matrix Scientific

073635-1G 073635-5G
102531-206EA 853.85 USD
102531-206 102531-208
3-Methoxyisonicotinic acid ≥95%
3-Methoxyisonicotinic acid

Hazard: Irritant

Formula: C₇H₇NO₃
MDL Number: MFCD07368621
CAS Number: 654663-32-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR