tert-Butyl-1-bromo-5,6-dihydroimidazo[1,5-a]pyrazine-7(8H)-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
tert-Butyl 1-bromo-5,6-dihydroimidazo[1,5-a]pyrazine-7(8H)-carboxylate
074551-250MG
074551-5G
102537-006EA
640
USD
102537-006
102537-010
tert-Butyl-1-bromo-5,6-dihydroimidazo[1,5-a]pyrazine-7(8H)-carboxylate ≥95%
tert-Butyl-1-bromo-5,6-dihydroimidazo[1,5-a]pyrazine-7(8H)-carboxylate
Hazard: Irritant
Formula:
C₁₁H₁₆BrN₃O₂ MW: 302.1700 g/mol |
MDL Number:
MFCD13152218 CAS Number: 1188265-64-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...