3-(N,N-Dimethylamino)phenylboronic acid hydrochloride ≥95%

Supplier: Matrix Scientific

074087-1G 074087-5G 074087-10G
102534-570EA 90.77 USD
102534-570 102534-572 102534-574
3-(N,N-Dimethylamino)phenylboronic acid hydrochloride ≥95%
3-(N,N-Dimethylamino)phenylboronic acid hydrochloride

Hazard: Irritant

Formula: C₈H₁₃BClNO₂
MW: 201.4583 g/mol
MDL Number: MFCD04112542
CAS Number: 1256355-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR