(R)-1-((Benzyloxy)carbonyl)piperidine-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

074061-1G 074061-5G
102534-430EA 93.85 USD
102534-430 102534-432
(R)-1-((Benzyloxy)carbonyl)piperidine-3-carboxylic acid ≥95%
(R)-1-((Benzyloxy)carbonyl)piperidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₇NO₄
MW: 263.2900 g/mol
MDL Number: MFCD04114972
CAS Number: 160706-62-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR