tert-Butyl ((2S,4S,5S)-5-amino-4-hydroxy-1,6-diphenylhexan-2-yl)carbamate ≥95%

Supplier: Matrix Scientific

074025-1G 074025-5G
102534-238EA 213.85 USD
102534-238 102534-240
tert-Butyl ((2S,4S,5S)-5-amino-4-hydroxy-1,6-diphenylhexan-2-yl)carbamate ≥95%
tert-Butyl ((2S,4S,5S)-5-amino-4-hydroxy-1,6-diphenylhexan-2-yl)carbamate

Hazard: Irritant

Formula: C₂₃H₃₂N₂O₃
MW: 384.5118 g/mol
MDL Number: MFCD09833420
CAS Number: 144163-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR