(6R,7R)-4-Methoxybenzyl-7-amino-3-(chloromethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: ACLE HCL, (6R,7R)-4-Methoxybenzyl 7-amino-3-(chloromethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate hydrochloride

073982-5G 073982-25G
102534-028EA 283.08 USD
102534-028 102833-664
(6R,7R)-4-Methoxybenzyl-7-amino-3-(chloromethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate hydrochloride ≥95%
(6R,7R)-4-Methoxybenzyl-7-amino-3-(chloromethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate hydrochloride

Hazard: Irritant

Formula: C₁₆H₁₈Cl₂N₂O₄S
MW: 405.2961 g/mol
MDL Number: MFCD00191255
CAS Number: 113479-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR