1-(3,3-Diphenyl-N-methylpropylamino)-2-methyl-2-propanol ≥95%

Supplier: Matrix Scientific

Synonyms: 1-[(3,3-Diphenylpropyl)(methyl)amino]-2-methyl-2-propanol, 1-[(3,3-Diphenylpropyl)methylamino]-2-methyl-2-propanol

073962-10G 073962-100G
102533-922EA 107.69 USD
102533-922 102533-924
1-(3,3-Diphenyl-N-methylpropylamino)-2-methyl-2-propanol ≥95%
1-(3,3-Diphenyl-N-methylpropylamino)-2-methyl-2-propanol

Hazard: Irritant

Formula: C₂₀H₂₇NO
MW: 297.43 g/mol
MDL Number: MFCD07782118
CAS Number: 100442-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR