(R)-Ethyl-3-((R)-2,2-dimethyl-1,3-dioxolan-4-yl)-2,2-difluoro-3-hydroxypropanoate ≥95%

Supplier: Matrix Scientific

073894-1G 073894-5G 073894-10G
102533-568EA 107.69 USD
102533-568 102533-570 102533-572
(R)-Ethyl-3-((R)-2,2-dimethyl-1,3-dioxolan-4-yl)-2,2-difluoro-3-hydroxypropanoate ≥95%
(R)-Ethyl-3-((R)-2,2-dimethyl-1,3-dioxolan-4-yl)-2,2-difluoro-3-hydroxypropanoate

Hazard: Irritant

Formula: C₁₀H₁₆F₂O₅
CAS Number: 95058-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR