N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine ≥90%

Supplier: Matrix Scientific

Synonyms: Benzyl(methoxymethyl)[(trimethylsilyl)methyl]amine

073881-5G 073881-25G
102533-494EA 73.85 USD
102533-494 102833-656
N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine ≥90%
N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine

Hazard: Irritant

Formula: C₁₃H₂₃NOSi
MW: 237.42 g/mol
MDL Number: MFCD00674005
CAS Number: 93102-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR