1-Benzenesulfonyl-1H-pyrrolo[2,3-b]pyridine-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

073873-250MG 073873-1G
102533-456EA 826.15 USD
102533-456 102533-458
1-Benzenesulfonyl-1H-pyrrolo[2,3-b]pyridine-2-carboxylic acid ≥95%
1-Benzenesulfonyl-1H-pyrrolo[2,3-b]pyridine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₀N₂O₄S
CAS Number: 189089-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR