Ethyl-1-methyl-1H-benzo[d][1,2,3]triazole-5-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 1-methyl-1H-benzo[d][1,2,3]triazole-5-carboxylate

073860-1G 073860-5G 073860-10G
102533-384EA 640 USD
102533-384 102533-386 102533-388
Ethyl-1-methyl-1H-benzo[d][1,2,3]triazole-5-carboxylate ≥95%
Ethyl-1-methyl-1H-benzo[d][1,2,3]triazole-5-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₁N₃O₂
MDL Number: MFCD04110946
CAS Number: 499785-52-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR