tert-Butyl ((5-bromopyridin-2-yl)methyl)carbamate ≥95%

Supplier: Matrix Scientific

073840-250MG 073840-1G 073840-5G
102533-270EA 640 USD
102533-270 102533-272 102533-274
tert-Butyl ((5-bromopyridin-2-yl)methyl)carbamate ≥95%
tert-Butyl ((5-bromopyridin-2-yl)methyl)carbamate

Hazard: Irritant

Formula: C₁₁H₁₅BrN₂O₂
MW: 287.1530 g/mol
MDL Number: MFCD11520868
CAS Number: 1188477-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR