tert-Butyl-6-bromo-4H-spiro[benzo[d][1,3]dioxine-2,4'-piperidine]-1'-carboxylate ≥95%

Supplier: Matrix Scientific

073791-250MG 073791-1G
102533-016EA 684.62 USD
102533-016 102533-018
tert-Butyl-6-bromo-4H-spiro[benzo[d][1,3]dioxine-2,4'-piperidine]-1'-carboxylate ≥95%
tert-Butyl-6-bromo-4H-spiro[benzo[d][1,3]dioxine-2,4'-piperidine]-1'-carboxylate

Hazard: Irritant

Formula: C₁₇H₂₂BrNO₄
CAS Number: 895525-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR