Spiro[piperidine-4,4'-pyrido[2,3-d][1,3]oxazin]-2'(1'H)-one ≥95%

Supplier: Matrix Scientific

073785-250MG 073785-1G
102531-986EA 826.15 USD
102531-986 102531-988
Spiro[piperidine-4,4'-pyrido[2,3-d][1,3]oxazin]-2'(1'H)-one ≥95%
Spiro[piperidine-4,4'-pyrido[2,3-d][1,3]oxazin]-2'(1'H)-one

Hazard: Irritant

Formula: C₁₁H₁₃N₃O₂
CAS Number: 753440-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR