tert-Butyl (2-(4-aminophenyl)-2-methylpropyl)carbamate ≥95%

Supplier: Matrix Scientific

073742-250MG 073742-1G
102531-764EA 826.15 USD
102531-764 102531-766
tert-Butyl (2-(4-aminophenyl)-2-methylpropyl)carbamate ≥95%
tert-Butyl (2-(4-aminophenyl)-2-methylpropyl)carbamate

Hazard: Irritant

Formula: C₁₅H₂₄N₂O₂
MDL Number: MFCD13195344
CAS Number: 180081-10-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR