Ethyl-4-chlorothieno[3,2-d]pyrimidine-2-carboxylate ≥95%

Supplier: Matrix Scientific

073730-250MG 073730-1G 073730-5G
102531-706EA 684.62 USD
102531-706 102531-708 102531-710
Ethyl-4-chlorothieno[3,2-d]pyrimidine-2-carboxylate ≥95%
Ethyl-4-chlorothieno[3,2-d]pyrimidine-2-carboxylate

Hazard: Irritant

Formula: C₉H₇ClN₂O₂S
CAS Number: 319442-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR