Ethyl-6-bromo-8-fluoroimidazo[1,2-a]pyridine-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 6-bromo-8-fluoroimidazo[1,2-a]pyridine-2-carboxylate

073706-250MG 073706-1G 073706-5G
102531-588EA 598.46 USD
102531-588 102531-590 102531-592
Ethyl-6-bromo-8-fluoroimidazo[1,2-a]pyridine-2-carboxylate ≥95%
Ethyl-6-bromo-8-fluoroimidazo[1,2-a]pyridine-2-carboxylate

Hazard: Irritant

Formula: C₁₀H₈BrFN₂O₂
MW: 287.09 g/mol
MDL Number: MFCD13195312
CAS Number: 1260763-32-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR