Ethyl-5-chloro-1H-pyrrolo[3,2-b]pyridine-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 5-chloro-1H-pyrrolo[3,2-b]pyridine-2-carboxylate

073695-250MG 073695-1G 073695-5G
102531-528EA 740 USD
102531-528 102531-530 102531-532
Ethyl-5-chloro-1H-pyrrolo[3,2-b]pyridine-2-carboxylate ≥95%
Ethyl-5-chloro-1H-pyrrolo[3,2-b]pyridine-2-carboxylate

Hazard: Irritant

Formula: C₁₀H₉ClN₂O₂
MW: 224.6437 g/mol
MDL Number: MFCD11109466
CAS Number: 800401-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR