4-Bromo-2-nitrotoluene ≥98%

Supplier: Matrix Scientific

073633-25G 073633-100G
102531-196EA 44.62 USD
102531-196 102531-198
4-Bromo-2-nitrotoluene ≥98%
4-Bromo-2-nitrotoluene

Hazard: Irritant

Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
MDL Number: MFCD00041243
CAS Number: 60956-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR