5-Fluoro-3-nitropyridin-2(1H)-one ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Fluoro-2-hydroxy-3-nitropyridine

073630-1G 073630-5G 073630-10G
102531-182EA 83.08 USD
102531-182 102531-184 102531-186
5-Fluoro-3-nitropyridin-2(1H)-one ≥95%
5-Fluoro-3-nitropyridin-2(1H)-one

Hazard: Irritant

Formula: C5H3FN2O3
MW: 158.09 g/mol
MDL Number: MFCD05662412
CAS Number: 136888-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR