3-(Phenyldiazenyl)pyridine-2,6-diamine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: Phenazopyridine hydrochloride, 3-Phenylazo-2,6-diaminopyridine hydrochloride

073629-1G 073629-5G 073629-10G
102531-176EA 72.31 USD
102531-176 102531-178 102531-180
3-(Phenyldiazenyl)pyridine-2,6-diamine hydrochloride ≥95%
3-(Phenyldiazenyl)pyridine-2,6-diamine hydrochloride

Hazard: Irritant

Formula: C11H12ClN5
MW: 249.70 g/mol
MDL Number: MFCD00035347
CAS Number: 136-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR